missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH5-07, MedChemExpress
Description
SH5-07 is a hydroxamic acid based Stat3 inhibitor with an IC50 of 3.9 μM in in vitro assay.
Specifications
Specifications
| Kemisk navn eller materiale | SH5-07 |
| CAS | 1456632-41-9 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C29H28F5N3O5S |
| Mængde | 50 mg |
| Opløselighedsinformation | DMSO : 50 mg/mL (79.92 mM; Need ultrasonic) |
| SMIL | O=C(NO)C1=CC=C(N(CC2=CC=C(C3CCCCC3)C=C2)C(CN(C)S(=O)(C4=C(F)C(F)=C(F)C(F)=C4F)=O)=O)C=C1 |
| Molekylvægt (g/mol) | 625.61 |
| Formel vægt | 625.61 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?