Learn More
Keystone Cleanroom Products Self-Sealing 1422A Tyvek™ Autoclave Bag with Steam Indicator, 0 to 6 in. wide
The Self-Sealing 1422A Tyvek™ Autoclave Bag with Steam Indicator, 0 to 6 in. wide is designed for the steam sterilization of pharmaceutical equipment change parts. These bags are closed via the convenient and robust self-sealing closure system.
11070.00 DKK - 27170.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Closure Type | Self-seal |
Color | White |
Disposable | Single-use |
For Use With (Application) | Steam Sterilization |
Product Code | Brand | Length (English) | Width (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Width (English) | Price | Quantity & Availability | |||||
17787867
![]() |
Keystone Cleanroom Products
BGT2A0205SS |
5 in. | 2 in. |
11070.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17767887
![]() |
Keystone Cleanroom Products
BGT2A0404SS |
4 in. | 4 in. |
13420.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17777887
![]() |
Keystone Cleanroom Products
BGT2A0409SS |
9 in. | 4 in. |
25155.00 DKK
Pack of 500 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17797887
![]() |
Keystone Cleanroom Products
BGT2A0416SS |
16 in. | 4 in. |
14760.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17737897
![]() |
Keystone Cleanroom Products
BGT2A0426SS |
26 in. | 4 in. |
15435.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17757897
![]() |
Keystone Cleanroom Products
BGT2A0428SS |
28 in. | 4 in. |
15935.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17767897
![]() |
Keystone Cleanroom Products
BGT2A0430SS |
30 in. | 4 in. |
16940.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17777907
![]() |
Keystone Cleanroom Products
BGT2A0608SS |
8 in. | 6 in. |
11405.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17707917
![]() |
Keystone Cleanroom Products
BGT2A0610SS |
10 in. | 6 in. |
26500.00 DKK
Pack of 500 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17717917
![]() |
Keystone Cleanroom Products
BGT2A0612SS |
12 in. | 6 in. |
27170.00 DKK
Pack of 500 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17737917
![]() |
Keystone Cleanroom Products
BGT2A0615SS |
15 in. | 6 in. |
14090.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17757917
![]() |
Keystone Cleanroom Products
BGT2A0616SS |
16 in. | 6 in. |
14760.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17777917
![]() |
Keystone Cleanroom Products
BGT2A0638SS |
38 in. | 6 in. |
22475.00 DKK
Pack of 250 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Integrated steam indicator
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in controlled cleanroom environment
- Eliminates need to cut material and the use of autoclave tape
- Self-sealing closure is fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Resists tearing and punctures
- Superior microbial barrier
- Certificates of conformance sent with shipment
Specifications
Autoclavable | |
White | |
Steam Sterilization | |
Tyvek | |
Autoclave Bag | |
Autoclave Bag | |
Autoclave Bag | |
Autoclave Bag | |
Autoclave Bag | |
Autoclave Bag |
Self-seal | |
Single-use | |
Steam Sterilization Indicator | |
Non-sterile | |
Yes | |
Autoclave Bag | |
Autoclave Bag | |
Autoclave Bag | |
Autoclave Bag |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Keystone Cleanroom Products Self-Sealing 1422A Tyvek™ Autoclave Bag with Steam Indicator, 0 to 6 in. wide