Learn More
3M™ Secure Click™ Reusable Full Face Respirator
The Secure Click™ Reusable Full Face Mask FF-800 Series helps protect against particulates and a wide variety of gases and vapors when used with approved Secure Click™ filters.
3390.00 DKK
Specifications
Color Black
Filter Type Secure Click™ Filters
View More Specs
Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17976640
View Documents
3M™
7100262627
Small
3390.00 DKK
Each
Estimated Shipment: 02-07-2025
Add to basket
17956640
View Documents
3M™
7100262628
Medium
3390.00 DKK
Each
Estimated Shipment: 02-07-2025
Add to basket
17966640
View Documents
3M™
7100262626
Large
3390.00 DKK
Each
Estimated Shipment: 02-07-2025
Add to basket
Description
Description
  • The quick, easy, one-touch positive pressure push button seal check helps provide improved confidence in respirator fit
  • Two dual-flow gas and vapor filters on each respirator combine to create the world's first quad flow gas and vapor filter system with four airflow paths providing easy breathing and comfort
  • Large lens for excellent field of vision, and treated with Scotchgard™ Protector for improved paint and stain resistance
  • Six-strap harness helps provide a secure comfortable fit
  • Speaking diaphragm is designed to help provide easy communication
  • Recommended applications include assembly, chemical clean-up, chemical handling, chipping, chiseling, cleaning, furnace operations, grinding, machining, masonry, painting, pouring/casting, sanding and sawing.
  • Available in three sizes
Specifications
Specifications
Black
Secure Click™ Filters
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Secure Click™ Reusable Full Face Respirator