Learn More
Lenz Laborglasintrumente Screwthread Tubes For Glassblowers
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01300045
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10048440
  • 1255.00 DKK / Pack of 10
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12216 and in addition to standard
  • With DIN thread (GL)
  • Material: DURAN™
Specifications
Specifications
40 mm
2.3 mm
45 GL
Duran Glass
10/Pk.