Få mere at vide
Scandium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 047199.06
| Mængde | 5g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Kemiske identifikatorer
| 13465-59-3 | |
| 284.67 | |
| APPHYFNIXVIIJR-UHFFFAOYSA-K | |
| 83495 | |
| [Sc+3].[Br-].[Br-].[Br-] |
| Br3Sc | |
| MFCD00016324 | |
| scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis | |
| scandium(3+) tribromide |
Tekniske data
| Scandium(III) bromide | |
| 13465-59-3 | |
| (metals basis) | |
| MFCD00016324 | |
| APPHYFNIXVIIJR-UHFFFAOYSA-K | |
| scandium(3+) tribromide | |
| 83495 | |
| 99.99% | |
| Hygroscopic |
| Ultra dry | |
| 5g | |
| Br3Sc | |
| scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis | |
| [Sc+3].[Br-].[Br-].[Br-] | |
| 284.67 | |
| 284.67 | |
| Ampule |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.