missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SBK1, GST Tag Recombinant Protein
Recombinant Protein
4245.00 DKK - 61400.00 DKK
Specifications
Accession Number | Q52WX2 |
---|---|
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 388228 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910293
|
Invitrogen™
A30518 |
10 ÎĽg |
4245.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17138785
|
Invitrogen™
A33367 |
1 mg |
61400.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17108745
|
Invitrogen™
A32877 |
100 ÎĽg |
19780.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SBK1 novel serine/threonine kinase or SH3 domain binding kinase 1 is structurally related to a Xenopus gastrula-specific protein kinase, Pk9.7 which possessing a consensus sequence for an SH3-binding domain from developing rat brain. SBK1 plays an important role in signal-transduction pathways related to the control of brain development. SBK1 may predict a broad cellular function, and its dysregulated in certain cancers suggests an involvement of the protein in the pathogenesis of human cancers. May be involved in signal-transduction pathways related to the control of brain development. Target protein is a serine/threonine protein kinase gene expressed predominantly in developing brain.Specifications
Q52WX2 | |
Kinase Assay | |
388228 | |
Human SBK1, GST Tag | |
Sbk1 | |
Unconjugated | |
Recombinant | |
Full length | |
Dry Ice | |
Full Length, Recombinant, Active |
See Label | |
50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
72.5 kDa | |
SBK; SBK1; Serine/threonine-protein kinase SBK1; SH3 domain binding kinase 1; SH3-binding domain kinase 1; SH3-binding kinase 1 | |
NKF1 Family | |
Human | |
GST-tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Insect cells | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human SBK1, GST Tag Recombinant Protein