missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human SBK1, GST Tag Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
Pakningsstørrelse:
10µg
Beskrivelse
SBK1 novel serine/threonine kinase or SH3 domain binding kinase 1 is structurally related to a Xenopus gastrula-specific protein kinase, Pk9.7 which possessing a consensus sequence for an SH3-binding domain from developing rat brain. SBK1 plays an important role in signal-transduction pathways related to the control of brain development. SBK1 may predict a broad cellular function, and its dysregulated in certain cancers suggests an involvement of the protein in the pathogenesis of human cancers. May be involved in signal-transduction pathways related to the control of brain development. Target protein is a serine/threonine protein kinase gene expressed predominantly in developing brain.

Tekniske data
Tekniske data
Adgangsnummer | Q52WX2 |
Koncentration | See Label |
Til brug med (applikation) | Kinase Assay |
Formulering | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gen-id (Entrez) | 388228 |
Molekylvægt (g/mol) | 72.5 kDa |
Navn | Human SBK1, GST Tag |
Mængde | 100 μg |
Gene Alias | SBK; SBK1; Serine/threonine-protein kinase SBK1; SH3 domain binding kinase 1; SH3-binding domain kinase 1; SH3-binding kinase 1 |
Fælles navn | Sbk1 |
Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Invitrogen™ Human SBK1, GST Tag Recombinant Protein >
Ser du en mulighed for forbedring?Del en indholdskorrektion