Få mere at vide
SB-743921 hydrochloride, MedChemExpress
Artikelnummer. 16494968
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16494968 10 mg
16461217 50 mg
16435746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16494968 Leverandør Medchem Express Leverandørnr. HY12069
MedChemExpress SB-743921 hydrochloride is a potent inhibitor of the mitotic kinesin KSP (Eg5), with a Ki of 0.1 nM.
SB-743921 hydrochloride is a potent inhibitor of the mitotic kinesin KSP (Eg5), with a Ki of 0.1 nM.
Tekniske data
Kemisk navn eller materiale SB-743921 hydrochloride
CAS 940929-33-9
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C31H34Cl2N2O3
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 100 mg/mL (180.66 mM) ∣H2O : 10 mg/mL (18.07 mM; ultrasonic and warming and heat to 60°C)
SMIL ClC1=CC2=C(C(C(CC3=CC=CC=C3)=C([C@@H](C(C)C)N(C(C4=CC=C(C=C4)C)=O)CCCN)O2)=O)C=C1.Cl
Molekylvægt (g/mol) 553.52
Formel vægt 553.52
Procent renhed 98.11%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-programmed cell death