missing translation for 'onlineSavingsMsg'
Få mere at vide

SB 239063, MedChemExpress

Artikelnummer. 16497107
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16497107 5 mg 5mg
16460707 50 mg -
16454388 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16497107 Leverandør Medchem Express Leverandørnr. HY11068

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress SB 239063 is a potent, selective and orally active p38 MAPK inhibitor, exhibits an IC50 of 44 nM for recombinant purified human p38α, with equipotent inhibitory activity against p38α and p38β. SB 239063 has no effect on p38γ or p38δ. With anti-asthma activity and also be used to enhance memory which is impaired due to aging or medical conditions, such as, AD.

SB 239063 is a potent, selective and orally active p38 MAPK inhibitor, exhibits an IC50 of 44 nM for recombinant purified human p38α, with equipotent inhibitory activity against p38α and p38β. SB 239063 has no effect on p38γ or p38δ. With anti-asthma activity and also be used to enhance memory which is impaired due to aging or medical conditions, such as, AD.

Tekniske data

Kemisk navn eller materiale SB 239063
CAS 193551-21-2
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C20H21FN4O2
Mængde 5 mg
Opløselighedsinformation DMSO : 33.33 mg/mL (90.47 mM; Need ultrasonic)
SMIL COC1=NC=CC(C2=C(N=CN2[C@H]3CC[C@H](O)CC3)C4=CC=C(F)C=C4)=N1
Molekylvægt (g/mol) 368.4
Formel vægt 368.4
Procent renhed 99.49%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.