Learn More
Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand:  Sartorius FT-3-207-185
189.18 DKK valid until 2024-02-28
Use promo code "21659" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10032301
  • 301.00 DKK / Pack of 100
Estimated Shipment: 15-03-2024
to see stock.
Add to basket
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
20 sec.
100/Pk.
185 mm
8 to 12 μm
1289
0.21 mm
Recommendations
Recommendations