Learn More
Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
84.25 DKK - 489.00 DKK
Specifications
Format Disc
Quantity 100/Pk.
Thickness (Metric) 0.21 mm
View More Specs
Products 11
Product Code Brand Diameter (Metric) Grade Particle Retention Price Quantity & Availability  
Product Code Brand Diameter (Metric) Grade Particle Retention Price Quantity & Availability  
10471232
View Documents Promotion Details
Sartorius
FT-3-208-070
70 mm 1290 3 to 5 μm
89.45 DKK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
10387781
View Documents Promotion Details
Sartorius
FT-3-208-055
55 mm 1290 3 to 5 μm
84.25 DKK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
10778772
View Documents Promotion Details
Sartorius
FT-3-207-240
240 mm 1289 8 to 12 μm
489.00 DKK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
10480272
View Documents Promotion Details
Sartorius
FT-3-207-125
125 mm 1289 8 to 12 μm
155.00 DKK
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
10032301
View Documents Dettagli della promozione
Sartorius
FT-3-207-185
185 mm 1289 8 to 12 μm
301.00 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-05-2024
to see stock.
Aggiungi al carrello
10205421
missing translation for 'viewDocuments' Dettagli della promozione
Sartorius
FT-3-207-110
110 mm 1289 8 to 12 μm
120.00 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-05-2024
to see stock.
Aggiungi al carrello
10133991
missing translation for 'viewDocuments' Dettagli della promozione
Sartorius
FT-3-207-070
70 mm 1289 8 to 12 μm
89.45 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 25-06-2024
to see stock.
Aggiungi al carrello
10767812
missing translation for 'viewDocuments' Dettagli della promozione
Sartorius
FT-3-207-090
90 mm 1289 8 to 12 μm
91.70 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 28-05-2024
to see stock.
Aggiungi al carrello
10490272
missing translation for 'viewDocuments' Dettagli della promozione
Sartorius
FT-3-207-150
150 mm 1289 8 to 12 μm
201.00 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-06-2024
to see stock.
Aggiungi al carrello
10226041
missing translation for 'viewDocuments' Dettagli della promozione
Sartorius
FT-3-207-055
55 mm 1289 8 to 12 μm
84.25 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 06-05-2024
to see stock.
Aggiungi al carrello
11761165
missing translation for 'viewDocuments' Dettagli della promozione
Sartorius
FT-3-207-050
50 mm 1289 8 to 12 μm
84.25 DKK
Pack of 100
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-06-2024
to see stock.
Aggiungi al carrello
Descrizione
Descrizione
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifica
Specifica
Disc
100/Pk.
0.21 mm
Videos
SDS
Documenti
Documenti
Certificati