missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartoclear Dynamics™ Lab V50

Sartoclear Dynamics™ Lab V50 Kit simplifies the process by fully eliminating the centrifugation step otherwise needed for clarification.

Brand:  Sartorius SDLV-0050-02E0-2

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 16423549

  • 3805.00 DKK / Pack of 24
Estimated Shipment: 08-07-2024
to see stock.
Add to basket

Description

Description

  • Enables clarification and filtration to be performed in a single step
  • Designed for the clarification and sterile filtration of up to 50 mL mammalian cell culture
  • Comprised of pouches of filter aid and vacuum filtration units
  • For a single-step mammalian cell culture harvesting
  • Gamma sterilized pouches
Specifications

Specifications

Cell Density: > 10 mio. cells/mL; Pore Size: 0.22 μm
2 g diatomaceous earth as filter aid per unit of filtration
Cell Culture Filtration
Filtration Kits
1 x 180E01---------2 (24 x Sartolab RF 50, 0.22 μm, PES, single packed sterile), 2 x SDLKG-01.0------2 (48 x pouches of filter aid, 1 g), 24 filtration units
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers