Få mere at vide
IBA Lifesciences SARS-CoV-2 Peptide KVGGNYNYL (HLA-A*0201)
Shop alle IBA Lifesciences produkter
Mængde:
1 mg
Beskrivelse
- SARS-CoV-2 peptide KVGGNYNYL (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Tekniske data
Tekniske data
| Renhed | >85% |
| Endotoksin niveau | Free |
| Format | Lyophilized Powder |
| Mængde | 1 mg |
| Forskningskategori | T cell stimulation |
For Research Use Only.