Få mere at vide
Sapitinib, MedChemExpress
Artikelnummer. 16471687
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16471687 50 mg
16458227 5 mg
16416096 100 mg
16415518 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16471687 Leverandør Medchem Express Leverandørnr. HY13050
MedChemExpress Sapitinib (AZD-8931) is a reversible, ATP competitive EGFR inhibitor of with IC50s of 4, 3 and 4 nM for EGFR, ErbB2 and ErbB3 in cells, respectively.
Sapitinib (AZD-8931) is a reversible, ATP competitive EGFR inhibitor of with IC50s of 4, 3 and 4 nM for EGFR, ErbB2 and ErbB3 in cells, respectively.
Tekniske data
Kemisk navn eller materiale Sapitinib
CAS 848942-61-0
Farve White
Renhedsgrad noter Research
Molekylær formel C23H25ClFN5O3
Mængde 50 mg
Synonym AZD-8931
Opløselighedsinformation DMSO : ≥ 33 mg/mL (69.63 mM)
SMIL O=C(CN1CCC(OC2=CC3=C(NC4=CC=CC(Cl)=C4F)N=CN=C3C=C2OC)CC1)NC
Molekylvægt (g/mol) 473.93
Formel vægt 473.93
Procent renhed 99.28%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease