missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SAMD14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
3675.00 DKK
Tekniske data
| Antigen | SAMD14 |
|---|---|
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Værtsarter | Rabbit |
Beskrivelse
SAMD14 Polyclonal specifically detects SAMD14 in Human samples. It is validated for Western Blot.Tekniske data
| SAMD14 | |
| Polyclonal | |
| Rabbit | |
| sterile alpha motif domain containing 14 | |
| SAMD14 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 201191 | |
| Synthetic peptides corresponding to SAMD14(sterile alpha motif domain containing 14) The peptide sequence was selected from the N terminal of SAMD14. Peptide sequence HKARAQLLAKGRRHRPSRSRLRDSASSAEDGEGSDGPGGKVTDGCGSPLH. | |
| Primary |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel