Få mere at vide
SAMD14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-70699
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SAMD14 Polyclonal specifically detects SAMD14 in Human samples. It is validated for Western Blot.
Tekniske data
| SAMD14 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| SAMD14 | |
| Synthetic peptides corresponding to SAMD14(sterile alpha motif domain containing 14) The peptide sequence was selected from the N terminal of SAMD14. Peptide sequence HKARAQLLAKGRRHRPSRSRLRDSASSAEDGEGSDGPGGKVTDGCGSPLH. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Pig: 92%. | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| sterile alpha motif domain containing 14 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 201191 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |