Få mere at vide
Salmeterol, TRC
Artikelnummer. 30007788
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007788 50 mg
17982045 10 mg
17933615 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007788 Leverandør Toronto Research Chemicals Leverandørnr. TRCS09010550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 89365-50-4
Molekylær formel C25 H37 N O4
Molekylvægt (g/mol) 415.57
Synonym 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-, 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-, (±)-, GR 33343X, Salmeterol
IUPAC navn 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol
SMIL OCc1cc(ccc1O)C(O)CNCCCCCCOCCCCc2ccccc2
Tekniske data
Kemisk navn eller materiale Salmeterol
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C25H37NO4/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2
Formel vægt 415.2723
Procent renhed >95%