Få mere at vide
Safinamide, MedChemExpress
Artikelnummer. 16444087
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
Pakningsstørrelse:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16444087 50 mg -
16418188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16444087 Leverandør Medchem Express Leverandørnr. HY70057
MedChemExpress Safinamide is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor (IC50=0.098 μM) over MAO-A (IC50=580 μM).
Safinamide also blocks sodium channels and modulates glutamate (Glu) release, showing a greater affinity at depolarized (IC50=8 μM) than at resting (IC50=262 μM) potentials. Safinamide has neuroprotective and neurorescuing effects and can be used for the study of parkinson disease, ischemia stroke etc.al.
Tekniske data
Kemisk navn eller materiale Safinamide
CAS 133865-89-1
Farve White
Renhedsgrad noter Research
Molekylær formel C17H19FN2O2
Mængde 50 mg
Synonym FCE 26743 EMD 1195686
Opløselighedsinformation DMSO : ≥ 100 mg/mL (330.75 mM)
SMIL O=C([C@@H](NCC1=CC=C(OCC2=CC(F)=CC=C2)C=C1)C)N
Molekylvægt (g/mol) 302.34
Formel vægt 302.34
Procent renhed 99.89%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation