missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
S100A/B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-538-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
S100A/B Polyclonal specifically detects S100A/B in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Tekniske data
| S100A/B | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:10-1:500 | |
| P23297, P56565 | |
| S100A1 | |
| Affinity purified S-100 protein isolated from cow brain. | |
| 0.1 mL | |
| Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| 6271 | |
| Bovine, Human | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| HistoZyme, containing less than 0.1% sodium azide with Sodium Azide | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody reacts with cow S-100 protein a and b. It shows strong crossreactivity with human S-100 a and b. In the brain, it labels glial and ependymal cells. Melanocytes and Langerhans cells of the skin stain positive with this antibody. | |
| Store at 4°C. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion