Få mere at vide
(+)-(S)-Tylophorine, TRC
Artikelnummer. 30449925
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30449925 10 mg
30449052 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30449925 Leverandør Toronto Research Chemicals Leverandørnr. TRCT89820010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 482-20-2
Molekylær formel C24H27NO4
Molekylvægt (g/mol) 393.48
Synonym (S)-9,11,12,13,13a,14-Hexahydro-2,3,6,7-tetramethoxydibenzo[f,h]pyrrolo[1,2-b]isoquinoline, NSC 717335, NSC 76387, NSTP 0G01, Tylophorin, (+)-Tylophorine, (13aS)-9,11,12,13,13a,14-Hexahydro-2,3,6,7-tetramethoxydibenzo[f,h]pyrrolo[1,2-b]isoquinoline, Tylophorine
IUPAC navn (13aS)-2,3,6,7-tetramethoxy-9,11,12,13,13a,14-hexahydrophenanthro[9,10-f]indolizine
SMIL [H][C@](N1C2)(CCC1)CC3=C2C(C=C(OC)C(OC)=C4)=C4C5=C3C=C(OC)C(OC)=C5
Tekniske data
Kemisk navn eller materiale (+)-(S)-Tylophorine
Mængde 10 mg
InChI formel InChI=1S/C24H27NO4/c1-26-21-9-16-15-8-14-6-5-7-25(14)13-20(15)19-12-24(29-4)23(28-3)11-18(19)17(16)10-22(21)27-2/h9-12,14H,5-8,13H2,1-4H3/t14-/m0/s1
Formel vægt 393.194