Få mere at vide
S-Methyl-L-cysteine sulfoxide, TRC
Artikelnummer. 30133285
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133285 1 g
17937154 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133285 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2995481G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6853-87-8
Molekylær formel C4H9NO3S
Molekylvægt (g/mol) 151.18
Synonym 3-(methylsulfinyl)-L-Alanine, Kale anemia factor NSC 226572
IUPAC navn (2R)-3-hydrosulfinyl-2-(methylamino)propanoic acid
SMIL O=C(O)[C@H](CS=O)NC
Tekniske data
Kemisk navn eller materiale S-Methyl-L-cysteine sulfoxide
Renhedsgrad noter HPLC
InChI formel InChI=1S/C4H8NO3S/c1-5-3(2-9-8)4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m0/s1
Mængde 1 g
Formel vægt 151.0303
Procent renhed >95%