Få mere at vide
(S)-(+)-Mepivacaine Hydrochloride, TRC
Artikelnummer. 30470446
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470446 25 mg
30473148 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30470446 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22507525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34333-71-6
Molekylær formel C15 H22 N2 O . H Cl
Molekylvægt (g/mol) 282.809
Synonym (2S)-N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride, (+)-Mepivacaine Hydrochloride, (+)-1-Methyl-2',6'-pipecoloxylidide, MonoHydrochloride, (+)-Mepivacaine Hydrochloride, (S)-N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride, Ripovacaine EP Impurity C
IUPAC navn (2S)-N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
SMIL Cl.CN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C
Tekniske data
Kemisk navn eller materiale (S)-(+)-Mepivacaine Hydrochloride
Mængde 25 mg
InChI formel InChI=1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H/t13-;/m0./s1
Formel vægt 282.15