Få mere at vide
(S)-Fluoxetine-d5 Hydrochloride, TRC
Artikelnummer. 17918329
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17918329 1 mg
17918739 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17918329 Leverandør Toronto Research Chemicals Leverandørnr. TRCF5970971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217680-98-2
Molekylær formel C17H14D5ClF3NO
Molekylvægt (g/mol) 350.82
Synonym (γS)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy](benzene-d5)propanamine Hydrochloride, (S)-N-Methyl-3-(4-trifluoromethylphenoxy)-3-(phenyl-d5)propylamine Hydrochloride, (+)-(S)-Fluoxetine-d5 Hydrochloride, (+)-Fluoxetine-d5 Hydrochloride;
SMIL [2H]C1=C([C@@H](OC2=CC=C(C(F)(F)F)C=C2)CCNC)C([2H])=C([2H])C([2H])=C1[2H].Cl
Tekniske data
Kemisk navn eller materiale (S)-Fluoxetine-d5 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H/t16-;/m0./s1/i2D,3D,4D,5D,6D;