Få mere at vide
(S)-(+)-2-Chlorophenylglycine Methyl Ester Tartrate Salt, TRC
Artikelnummer. 30134504
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134504 1 g
30138603 5 g
30009141 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134504 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4242701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 141109-15-1
Molekylær formel C9 H10 Cl N O2 . C4 H6 O6
Molekylvægt (g/mol) 349.72
Synonym Benzeneacetic acid, α-amino-2-chloro-, methyl ester, (αS)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Benzeneacetic acid, α-amino-2-chloro-, methyl ester, (+)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1)
IUPAC navn (2R,3R)-2,3-dihydroxybutanedioic acid;methyl (2S)-2-amino-2-(2-chlorophenyl)acetate
SMIL COC(=O)[C@@H](N)c1ccccc1Cl.O[C@H]([C@@H](O)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale (S)-(+)-2-Chlorophenylglycine Methyl Ester Tartrate Salt
Mængde 1 g
InChI formel InChI=1S/C9H10ClNO2.C4H6O6/c1-13-9(12)8(11)6-4-2-3-5-7(6)10;5-1(3(7)8)2(6)4(9)10/h2-5,8H,11H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t8-;1-,2-/m01/s1
Formel vægt 349.0564