| Synonym |
4H-1-Benzopyran-4-one, 3-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-, Flavone, 3,3',4',5,5',7-hexahydroxy-, (6-O-α-L-rhamnosyl-β-D-glucoside) (7CI), Ilixanthin (6CI), Rutin (8CI), 3-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, 3,3',4',5,7-Pentahydroxyflavone 3-O-rutinoside, 3,3',4',5,7-Pentahydroxyflavone 3-rutinoside, 3-O-Rutinosylquercetin, 3-Rutinosylquercetin, 5,7,3',4'-Tetrahydroxyflavonol-3-O-rutinoside, Birutan, C.I. 75730, Eldrin, Globulariacitrin, Globularicitrin, Ilixathin, Melin, Myrticalorin, Myrticolorin, Myticolorin, NSC 9220, Novarrutina, Osyritin, Osyritrin, Oxyritin, Paliuroside, Phytomelin, Quercetin 3-(6-O-α-L-rhamnopyranosyl-β-D-glucopyranoside), Quercetin 3-O-rutinoside, Quercetin 3-O-α-L-rhamnopyranosyl-(1→6)-β-D-glucopyranoside, Quercetin 3-O-α-L-rhamnopyranosyl-β-D-glucopyranoside, Quercetin 3-O-α-rhamnopyranosyl(1' '→6')-β-D-glucopyranoside, Quercetin 3-O-α-rhamnopyranosyl(1→6)-β-glucopyranoside, Quercetin 3-O-α-rhamnopyranosyl-(1' ' '→6' ')-β-glucopyranoside, Quercetin 3-O-β-D-(6' '-O-α-L-rhamnopyranosyl)glucopyranoside, Quercetin 3-O-β-D-rutinoside, Quercetin 3-O-β-rutinoside, Quercetin 3-rhamnoglucoside, Quercetin 3-rutinoside, Quercetin 3-β-rutinoside, Quercetin 6-O-α-L-rhamnosyl-β-D-glucoside, Quercetin rutinoside, Rutabion, Rutine, Rutinic acid, Rutosid, Rutoside, Rutozid, Sophorin, Tanrutin, Violaquercetrin, Violaquercitrin, Yunxianggan |
| IUPAC navn |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |