missing translation for 'onlineSavingsMsg'
Få mere at vide

Rucaparib phosphate, MedChemExpress

Artikelnummer. 16484068
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
Pakningsstørrelse:
200mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16484068 10 mg -
16445046 100 mg -
16440427 50 mg -
16406777 5 mg -
16490996 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16484068 Leverandør Medchem Express Leverandørnr. HY10617

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Rucaparib (AG014699) phosphate is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib phosphate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib phosphate has the potential for castration-resistant prostate cancer (CRPC) research.

Rucaparib (AG014699) phosphate is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib phosphate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib phosphate has the potential for castration-resistant prostate cancer (CRPC) research.

Tekniske data

Kemisk navn eller materiale Rucaparib phosphate
CAS 459868-92-9
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C19H21FN3O5P
Mængde 10 mg
Synonym AG-014699 phosphatePF-01367338 phosphate
Opløselighedsinformation DMSO : ≥ 33 mg/mL (78.32 mM) ∣H2O : 5 mg/mL (11.87 mM; ultrasonic and warming and heat to 60°C)
SMIL O=P(O)(O)O.FC1=CC2=C3C(CCNC2=O)=C(C4=CC=C(CNC)C=C4)NC3=C1
Molekylvægt (g/mol) 421.36
Formel vægt 421.36
Procent renhed 98.77%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.