Learn More
uvex Rubiflex NB60SZ Chemical Protection Gloves
Full NBR-coating reinforced with cotton interlock supporting material for applications that require protection against chemicals and mechanical risks. Elastic on the glove cuff provides additional protection by preventing hazards from entering the glove.
338.00 DKK - 3370.00 DKK
Specifications
Cuff Material | Elastic |
---|---|
Applications | Laboratory |
Coating Material | NBR |
Color | Green |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17200411
|
uvex
6018060 |
9 |
3370.00 DKK
10 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17290242
|
uvex
9893061 |
10 |
3370.00 DKK
10 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17210411
|
uvex
6018062 |
11 |
338.00 DKK
1 pair |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Extended length chemical protection glove with reinforced cotton interlock lining
- Cotton liner helps absorb water vapor
- Additional elastic collar at gauntlet end
- Resistant to many chemicals, acids, alkalis, mineral oils, and solvents
- Suitable for applications that require protection against chemicals and mechanical risks (ex. heat)
Specifications
Elastic | |
NBR | |
Yes | |
Chemical-resistant Gloves | |
Type B | |
Chemical Handling, Drainage Construction, and Municipal Cleaning | |
Gauntlet | |
Rubiflex NB60SZ |
Laboratory | |
Green | |
Nitrile | |
2121X | |
JKOPT | |
Unisex | |
Cotton | |
60 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.