Learn More
Gilac™ Round Selective Sorting Dustbin
Round Selective Sorting Dustbin is great as a highly durable recycling bin. Reinforced bottom with anti-wear skirt.
Brand:  Gilac™ G613236-1
 View more versions of this product
Product Code. 17978391
  • 347.00 DKK / Each
Only null left
Estimated Shipment: 27-08-2025
Description
  • Round dustbin great as a recycling bin.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Specifications
50 L
24 in.
Green
Round
18.3 in.
Dust Bin
610 mm
13.2 gal.
465 mm
Not Autoclavable
HDPE
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gilac™ Round Selective Sorting Dustbin > Capacity: 50 L, Color: Green