Learn More
Gilac™ Round Selective Sorting Dustbin
Round Selective Sorting Dustbin is great as a highly durable recycling bin. Reinforced bottom with anti-wear skirt.
Brand:  Gilac™ G612028-1
 View more versions of this product
Product Code. 17968831
  • 503.00 DKK / Each
Only null left
Estimated Shipment: 21-07-2025
Description
  • Round dustbin great as a recycling bin.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Specifications
75 L
26.4 in.
Blue
Round
19.1 in.
Dust Bin
670 mm
19.8 gal.
485 mm
Not Autoclavable
HDPE
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gilac™ Round Selective Sorting Dustbin > Capacity: 75 L, Color: Blue