Få mere at vide
Rosiptor, MedChemExpress
Artikelnummer. 16445186
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16445186 100 mg -
16440587 50 mg -
16424258 10 mg 10mg
16436977 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16445186 Leverandør Medchem Express Leverandørnr. HY109011
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
MedChemExpress Rosiptor (AQX-1125) is a selective and orally active phosphatase SHIP1 activator with anti-inflammatory effects. Rosiptor (AQX-1125) inhibits Akt phosphorylation, inflammatory mediator production and leukocyte chemotaxis in vitro.
Rosiptor (AQX-1125) is a selective and orally active phosphatase SHIP1 activator with anti-inflammatory effects. Rosiptor (AQX-1125) inhibits Akt phosphorylation, inflammatory mediator production and leukocyte chemotaxis in vitro.
Tekniske data
Kemisk navn eller materiale Rosiptor
CAS 782487-28-9
Farve White
Renhedsgrad noter Research
Molekylær formel C20H35NO2
Mængde 100 mg
Synonym AQX-1125
Opløselighedsinformation DMSO : 150 mg/mL (466.56 mM; Need ultrasonic and warming)
SMIL C[C@@]([C@H]1CO)(CC[C@H](O)C1)[C@](CC[C@@]2(C)[C@@]3([H])CCC2=C)([H])[C@H]3CN
Molekylvægt (g/mol) 321.5
Formel vægt 321.5
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease