Få mere at vide
Rogaratinib, MedChemExpress
Artikelnummer. 16455487
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16455487 5 mg
16454206 100 mg
16439446 50 mg
16432748 10 mg
16400499 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16455487 Leverandør Medchem Express Leverandørnr. HY100019
MedChemExpress Rogaratinib (BAY1163877) is a potent and selective fibroblast growth factor receptor (FGFR) inhibitor.
Rogaratinib (BAY1163877) is a potent and selective fibroblast growth factor receptor (FGFR) inhibitor.
Tekniske data
Kemisk navn eller materiale Rogaratinib
CAS 1443530-05-9
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C23H26N6O3S
Mængde 5 mg
Synonym BAY1163877
Opløselighedsinformation DMSO : 5 mg/mL (10.72 mM; Need ultrasonic)
SMIL O=C1NCCN(CC2=C(COC)C(C3=CC4=CC(C)=CC(OC)=C4S3)=C5C(N)=NC=NN52)C1
Molekylvægt (g/mol) 466.56
Formel vægt 466.56
Procent renhed 98.0%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-Kinase/protease