Få mere at vide
Roflumilast, MedChemExpress
Artikelnummer. 16479117
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16479117 5 mg 5mg
16462627 50 mg -
16436538 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16479117 Leverandør Medchem Express Leverandørnr. HY15455
MedChemExpress Roflumilast is a selective PDE4 inhibitor with IC50s of 0.7, 0.9, 0.7, and 0.2 nM for PDE4A1, PDEA4, PDEB1, and PDEB2, respectively, without affecting PDE1, PDE2, PDE3 or PDE5 isoenzymes from various cells.
Roflumilast is a selective PDE4 inhibitor with IC50s of 0.7, 0.9, 0.7, and 0.2 nM for PDE4A1, PDEA4, PDEB1, and PDEB2, respectively, without affecting PDE1, PDE2, PDE3 or PDE5 isoenzymes from various cells.
Tekniske data
Kemisk navn eller materiale Roflumilast
CAS 162401-32-3
Farve White
Renhedsgrad noter Research
Molekylær formel C17H14Cl2F2N2O3
Mængde 5 mg
Opløselighedsinformation DMSO : ≥ 50 mg/mL (124.00 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(NC1=C(Cl)C=NC=C1Cl)C2=CC=C(OC(F)F)C(OCC3CC3)=C2
Molekylvægt (g/mol) 403.21
Formel vægt 403.21
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-anti-virus