Få mere at vide
Rituximab Mouse, FITC, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
Brand: ACROBiosystems RIB-FY35c-200ug
Beskrivelse
Rituximab Monoclonal antibody specifically detects Rituximab.Tekniske data
| Rituximab | |
| FITC | |
| 200 μg | |
| Primary | |
| IgG1 |
| Monoclonal | |
| Hybridoma | |
| RUO | |
| -20°C, aviod light |