Få mere at vide
Rilapladib, MedChemExpress
Artikelnummer. 16464756
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16464756 100 mg 100mg
16463678 10 mg -
16460067 50 mg 50mg
16436417 5 mg -
16422826 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16464756 Leverandør Medchem Express Leverandørnr. HY102004
MedChemExpress Rilapladib (SB 659032) is a selective Lp-PLA2 (lipoprotein-associated phospholipase A2) inhibitor with an IC50 of 230 pM. Rilapladib (SB 659032) is also a PAFR (Platelet Activating Factor Receptor) antagonist.
Rilapladib (SB 659032) is a selective Lp-PLA2 (lipoprotein-associated phospholipase A2) inhibitor with an IC50 of 230 pM. Rilapladib (SB 659032) is also a PAFR (Platelet Activating Factor Receptor) antagonist.
Tekniske data
Kemisk navn eller materiale Rilapladib
CAS 412950-08-4
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C40H38F5N3O3S
Mængde 100 mg
Synonym SB 659032
Opløselighedsinformation DMSO : 86.67 mg/mL (117.79 mM; Need ultrasonic)
SMIL O=C(N(C1CCN(CCOC)CC1)CC2=CC=C(C3=CC=C(C(F)(F)F)C=C3)C=C2)CN4C(SCC5=CC=CC(F)=C5F)=CC(C6=C4C=CC=C6)=O
Molekylvægt (g/mol) 735.81
Formel vægt 735.81
Procent renhed 99.71%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation