Learn More
Brady™ Rigid DuraSleeve™ Vinyl Wire Marking Inserts for M710
Rigid DuraSleeve™ Wire Marking Inserts for use with Brady Printer M710.
294.00 DKK - 414.00 DKK
Specifications
Adhesive Type | Non-adhesive |
---|---|
Finish | Gloss |
Format | Tape |
Material | Vinyl |
Product Type | Wire and Cable Markers |
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
17828024
|
Brady™
311994 |
White | 4.4 x 10 mm |
294.00 DKK
Pack of 500 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17888094
|
Brady™
311997 |
White | 4.4 x 12 mm |
343.00 DKK
Pack of 500 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
17847944
|
Brady™
312000 |
White | 4.4 x 16 mm |
357.00 DKK
Pack of 500 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
17858024
|
Brady™
312002 |
White | 4.4 x 23 mm |
372.00 DKK
Pack of 500 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17857944
|
Brady™
312005 |
White | 4.4 x 30 mm |
414.00 DKK
Pack of 500 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
17858094
|
Brady™
311995 |
Yellow | 4.4 x 10 mm |
294.00 DKK
Pack of 500 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17887854
|
Brady™
311998 |
Yellow | 4.4 x 12 mm |
343.00 DKK
Pack of 500 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17868094
|
Brady™
312003 |
Yellow | 4.4 x 23 mm |
372.00 DKK
Pack of 500 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
Description
- Brady™ DuraSleeve™ printable inserts create a durable legend
- Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
- Legend can be changed before or after termination
Specifications
Non-adhesive | |
Tape | |
Wire and Cable Markers | |
Data and Telecommunications Labeling, Wire and Cable Labeling | |
1 Label | |
-40°F to +158°F | |
0.0098 in. | |
0.17 in. |
Gloss | |
Vinyl | |
Non-sterile | |
M710, BMP71 | |
500 Labels | |
-40°C to +70°C | |
0.249 mm |