missing translation for 'onlineSavingsMsg'
Få mere at vide

Rifamycin SV sodium, 98+%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63095.03

Mængde 1g

 Se flere versioner af dette produkt

Produktkode 15475359

  • 768.67 DKK / 1g

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Antibiotic effective against mycobacteria. Can also inhibit DNA polymeraseRifamycin SV sodium is an antimicrobial agent used to inhibit the DNA polymerase and RNA polymerase in biochemistry. It is used for the treatment of HIV-related Tuberculosis, traveler’s diarrhea (TD), clostridium difficile-associated disease (CDAD) and rheumatoid knee synovitis. It is utilized to study potential genes like rif15 and rif16 during the conversion of rifamycin SV into rifamycin B.

Solubility
Soluble in water, alcohol and dimethyl sulfoxide.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Tekniske data

Kemiske identifikatorer

14897-39-3
720.77
YVOFSHPIJOYKSH-DITFEXNXSA-N
sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
C37H47NNaO12
MFCD09752825
124219641
[Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O

Tekniske data

Rifamycin SV sodium
Orange to Red
C37H47NNaO12
14,8218
YVOFSHPIJOYKSH-DITFEXNXSA-N
sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
124219641
≥98%
14897-39-3
1g
MFCD09752825
Soluble in water,alcohol and dimethyl sulfoxide.
[Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O
720.77
719.45
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.