Få mere at vide
RG13022, MedChemExpress
Artikelnummer. 16466087
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16466087 5 mg
16464606 100 mg
16409896 50 mg
16403368 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16466087 Leverandør Medchem Express Leverandørnr. HY101429
MedChemExpress RG13022 is a tyrosine kinase inhibitor; inhibits the autophosphorylation reaction of the EGF receptor with an IC50 of 4 μM.
RG13022 is a tyrosine kinase inhibitor
Tekniske data
Kemisk navn eller materiale RG13022
CAS 136831-48-6
Farve White
Renhedsgrad noter Research
Molekylær formel C16H14N2O2
Mængde 5 mg
Synonym Tyrphostin RG13022
Opløselighedsinformation DMSO : 100 mg/mL (375.53 mM; Need ultrasonic)
SMIL N#C/C(C1=CC=CN=C1)=C/C2=CC=C(OC)C(OC)=C2
Molekylvægt (g/mol) 266.29
Formel vægt 266.29
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation