Få mere at vide
Retinyl Retinoate, TRC
Artikelnummer. 30414120
Skift visning
Mængde:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414120 250 mg
30427552 25 mg
30409366 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30414120 Leverandør Toronto Research Chemicals Leverandørnr. TRCR275555250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15498-86-9
Molekylær formel C40H56O2
Molekylvægt (g/mol) 568.89
Synonym Retinoic Acid Retin-15-yl Ester, Retinoic Acid Ester with Retinol
IUPAC navn (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoate
SMIL O=C(OC/C=C(C)/C=C/C=C(C)/C=C/C1=C(C)CCCC1(C)C)/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C
Tekniske data
Kemisk navn eller materiale Retinyl Retinoate
Mængde 250 mg
InChI formel InChI=1S/C40H56O2/c1-30(21-23-36-34(5)19-13-26-39(36,7)8)15-11-17-32(3)25-28-42-38(41)29-33(4)18-12-16-31(2)22-24-37-35(6)20-14-27-40(37,9)10/h11-12,15-18,21-25,29H,13-14,19-20,26-28H2,1-10H3/b17-11+,18-12+,23-21+,24-22+,30-15+,31-16+,32-25+,33-29+
Formel vægt 568.43