Få mere at vide
rel-(1R,3S,5S)-8-Methyl-8-azabicyclo[3,2,1]oct-6-en-yl-2-hydroxy-2,2-dithiophen-2-yl acetate, TRC
Artikelnummer. 30465448
Skift visning
Mængde:
1 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465448 100 mg
30468490 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30465448 Leverandør Toronto Research Chemicals Leverandørnr. TRCM288620100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 136310-66-2
Molekylær formel C18 H19 N O3 S2
Molekylvægt (g/mol) 361.48
Synonym 2-Thiopheneacetic acid, α-hydroxy-α-2-thienyl-, 8-methyl-8-azabicyclo[3.2.1]oct-6-en-3-yl ester, endo-, Di(2-thienyl)glycolic acid tropenol ester, 2-Thiopheneacetic acid, α-hydroxy-α-2-thienyl-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-6-en-3-yl ester, Tiotropium Bromide Monohydrate Imp D (EP)
IUPAC navn [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]oct-6-en-3-yl] 2-hydroxy-2,2-dithiophen-2-ylacetate
SMIL CN1[C@@H]2C[C@H](C[C@H]1C=C2)OC(=O)C(O)(c3cccs3)c4cccs4
Tekniske data
Kemisk navn eller materiale (1R,3s,5S)-8-Methyl-8-azabicyclo[3.2.1]-oct-6-en-3-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate
Mængde 100 mg
InChI formel InChI=1S/C18H19NO3S2/c1-19-12-6-7-13(19)11-14(10-12)22-17(20)18(21,15-4-2-8-23-15)16-5-3-9-24-16/h2-9,12-14,21H,10-11H2,1H3/t12-,13+,14+
Formel vægt 361.0806