Learn More
Gilac™ Rectangular Dough Dividing Container
Rectangular Dough Dividing Container is shock-resistant, highly flexible, and resistant to changes in temperature. Nestable and stackable.
146.00 DKK - 148.00 DKK
Specifications
Type | Bin Divider |
---|---|
Material | HDPE |
Color | White |
Shape | Rectangular |
Certifications/Compliance | Food Contact Compliance |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15829572
|
Gilac™
G155921 |
13 L |
146.00 DKK
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
||||||
15125326
|
Gilac™
G155421 |
20 L |
148.00 DKK
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
Description
- Rectangular dough dividing container
- Highly resistant to changes in temperature
- Highly flexible and shock-resistant
- Compatible with racking trolleys
Specifications
Bin Divider | |
White | |
Food Contact Compliance | |
Stackable, Nestable | |
-40°C to +90°C |
HDPE | |
Rectangular | |
Rectangular | |
-40°F to +194°F |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Gilac™ Rectangular Dough Dividing Container