Learn More
Gilac™ Rectangular Dough Dividing Container
Rectangular Dough Dividing Container is shock-resistant, highly flexible, and resistant to changes in temperature. Nestable and stackable.
Brand: Gilac™ G155921
This item is not returnable.
View return policy
Description
- Rectangular dough dividing container
- Highly resistant to changes in temperature
- Highly flexible and shock-resistant
- Compatible with racking trolleys
Specifications
Bin Divider | |
White | |
3.4 gal. | |
Food Contact Compliance | |
GILAC Racking Trolley | |
Stackable, Nestable | |
23.6 in. | |
600 mm | |
15.7 in. | |
400 mm | |
77 mm | |
3 in. | |
0.752 kg | |
-40°C to +90°C |
HDPE | |
Rectangular | |
13 L | |
Rectangular | |
Food Storage | |
600 x 400 x 82 mm | |
19.7 in. | |
500 mm | |
11.8 in. | |
300 mm | |
82 mm | |
1.658 lb. | |
-40°F to +194°F |
Product Content Correction
Product Title
Gilac™ Rectangular Dough Dividing Container > Rectangular Dough Dividing Container; Capacity: 13 L
Spot an opportunity for improvement?Share a Content Correction