missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae GTP-binding protein YPT1 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) GTP-binding protein YPT1, was expressed in the hosts and tags indicated.

2615.00 DKK - 29485.00 DKK

Specifications

Accession Number NP_116615.1
Gene ID (Entrez) 850505
Name S. cerevisiae GTP-binding protein YPT1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16078320
View Documents
enQuireBio™
QP7341-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16018330
View Documents
enQuireBio™
QP7341-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068320
View Documents
enQuireBio™
QP7341-ye-100ug
100 ÎĽg
7930.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16098320
View Documents
enQuireBio™
QP7341-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16008330
View Documents
enQuireBio™
QP7341-ye-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16088320
View Documents
enQuireBio™
QP7341-ye-1mg
1 mg
29485.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_116615.1
S. cerevisiae GTP-binding protein YPT1 Protein
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
850505
Research Use Only
YPT1
Recombinant Protein
Yeast
MNSEYDYLFK LLLIGNSGVG KSCLLLRFSD DTYTNDYIST IGVDFKIKTV ELDGKTVKLQ IWDTAGQERF RTITSSYYRG SHGIIIVYDV TDQESFNGVK MWLQEIDRYA TSTVLKLLVG NKCDLKDKRV VEYDVAKEFA DANKMPFLET SALDSTNVED AFLTMARQIK ESMSQQNLNE TTQKKEDKGN VNLKGQSLTN TGGGCC
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant S. cerevisiae GTP-binding protein YPT1 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.