missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae GTP-binding protein YPT1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) GTP-binding protein YPT1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7341-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116615.1 | |
S. cerevisiae GTP-binding protein YPT1 Protein | |
Research Use Only | |
YPT1 | |
Recombinant Protein | |
Yeast | |
MNSEYDYLFK LLLIGNSGVG KSCLLLRFSD DTYTNDYIST IGVDFKIKTV ELDGKTVKLQ IWDTAGQERF RTITSSYYRG SHGIIIVYDV TDQESFNGVK MWLQEIDRYA TSTVLKLLVG NKCDLKDKRV VEYDVAKEFA DANKMPFLET SALDSTNVED AFLTMARQIK ESMSQQNLNE TTQKKEDKGN VNLKGQSLTN TGGGCC | |
Greater than 90% as determined by SDS-PAGE. |
850505 | |
50 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |