missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Lysyl endopeptidase Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Lysyl endopeptidase, was expressed in the hosts and tags indicated. option, ab
Brand: enQuireBio™ QP8146-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_252864.1 | |
P. aeruginosa Lysyl endopeptidase Protein | |
Research Use Only | |
prpL | |
Recombinant Protein | |
E. coli | |
AGYRDGFGAS GSCEVDAVCA TQSGTRAYDN ATAAVAKMVF TSSADGGSYI CTGTLLNNGN SPKRQLFWSA AHCIEDQATA ATLQTIWFYN TTQCYGDAST INQSVTVLTG GANILHRDAK RDTLLLELKR TPPAGVFYQG WSATPIANGS LGHDIHHPRG DAKKYSQGNV SAVGVTYDGH TALTRVDWPS AVVEGGSSGS GLLTVAGDGS YQLRGGLYGG PSYCGAPTSQ RNDYFSDFSG VYSQISRYFA P | |
Greater than 80% as determined by SDS-PAGE. |
880208 | |
50 μg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |