missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
1820.00 DKK - 19665.00 DKK
Specifications
Accession Number | NP_064295.2 |
---|---|
Gene ID (Entrez) | 56720 |
Name | Tryptophan 2,3-dioxygenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048281
|
enQuireBio™
QP9425-ye-10ug |
10 ÎĽg | N/A | ||||||
16088281
|
enQuireBio™
QP9425-ye-50ug |
50 ÎĽg | N/A | ||||||
16038281
|
enQuireBio™
QP9425-ye-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16068281
|
enQuireBio™
QP9425-ye-200ug |
200 ÎĽg | N/A | ||||||
16078281
|
enQuireBio™
QP9425-ye-500ug |
500 ÎĽg |
15875.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16058281
|
enQuireBio™
QP9425-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_064295.2 | |
Tryptophan 2,3-dioxygenase Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56720 | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
Yeast | |
MSGCPFAGNS VGYTLKNVSM EDNEEDRAQT GVNRASKGGL IYGNYLQLEK ILNAQELQSE VKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VIARMHRVVV IFKLLVQQFS VLETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQS LRVPYNRKHY RDNFGGDYNE LLLKSEQEQT LLQLVEAWLE RTPGLEPNGF NFWGKFEKNI LKGLEEEFLR IQAKTDSEEK EEQMAEFRKQ KEVLLCLFDE KRHDYLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDTL MTKWRYNHVC MVHRMLGTKA GTGGSSGYHY LRSTVSDRYK VFVDLFNLST YLVPRHWVPK MNPIIHKFLY TAEYSDSSYF SSDESD | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Tryptophan 2,3-dioxygenase Protein