missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9425-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_064295.2 | |
Tryptophan 2,3-dioxygenase Protein | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
Yeast | |
MSGCPFAGNS VGYTLKNVSM EDNEEDRAQT GVNRASKGGL IYGNYLQLEK ILNAQELQSE VKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VIARMHRVVV IFKLLVQQFS VLETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQS LRVPYNRKHY RDNFGGDYNE LLLKSEQEQT LLQLVEAWLE RTPGLEPNGF NFWGKFEKNI LKGLEEEFLR IQAKTDSEEK EEQMAEFRKQ KEVLLCLFDE KRHDYLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDTL MTKWRYNHVC MVHRMLGTKA GTGGSSGYHY LRSTVSDRYK VFVDLFNLST YLVPRHWVPK MNPIIHKFLY TAEYSDSSYF SSDESD | |
Greater than 90% as determined by SDS-PAGE. |
56720 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Tryptophan 2,3-dioxygenase Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction