missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MKK4 / MEK4 / MAP2K4 Protein
A DNA sequence encoding the Mus musculus (Mouse) MKK4 / MEK4 / MAP2K4, was expressed in the hosts and tags indicated.
1820.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_033183.1 |
---|---|
Gene ID (Entrez) | 26398 |
Name | MKK4 / MEK4 / MAP2K4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082570
|
enQuireBio™
QP6337-ec-10ug |
10 ÎĽg | N/A | ||||||
16022580
|
enQuireBio™
QP6337-ec-50ug |
50 ÎĽg | N/A | ||||||
16072570
|
enQuireBio™
QP6337-ec-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16002580
|
enQuireBio™
QP6337-ec-200ug |
200 ÎĽg | N/A | ||||||
16012580
|
enQuireBio™
QP6337-ec-500ug |
500 ÎĽg | N/A | ||||||
16092570
|
enQuireBio™
QP6337-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_033183.1 | |
MKK4 / MEK4 / MAP2K4 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26398 | |
Research Use Only | |
Map2k4 | |
Recombinant Protein | |
E. coli | |
AAPSPSGGGG SGGGGGTPGP IGPPASGHPA VSSMQGKRKA LKLNFANPPV KSTARFTLNP NTTGVQNPHI ERLRTHSIES SGKLKISPEQ HWDFTAEDLK DLGEIGRGAY GSVNKMVHKP SGQIMAVKRI RSTVDEKEQK QLLMDLDVVM RSSDCPYIVQ FYGALFREGD CWICMELMST SFDKFYKYVY SVLDDVIPEE ILGKITLATV KALNHLKENL KIIHRDIKPS NILLDRSGNI KLCDFGISGQ LVDSIAKTRD AGCRPYMAPE RIDPSASRQG YDVRSDVWSL GITLYELATG RFPYPKWNSV FDQLTQVVKG DPPQLSNSEE REFSPSFINF VNLCLTKDES KRPKYKELLK HPFILMYEER TVEVACYVCK ILDQMPATPS SPMYVD | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse MKK4 / MEK4 / MAP2K4 Protein