missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MKK4 / MEK4 / MAP2K4 Protein
A DNA sequence encoding the Mus musculus (Mouse) MKK4 / MEK4 / MAP2K4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6337-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_033183.1 | |
MKK4 / MEK4 / MAP2K4 Protein | |
Research Use Only | |
Map2k4 | |
Recombinant Protein | |
E. coli | |
AAPSPSGGGG SGGGGGTPGP IGPPASGHPA VSSMQGKRKA LKLNFANPPV KSTARFTLNP NTTGVQNPHI ERLRTHSIES SGKLKISPEQ HWDFTAEDLK DLGEIGRGAY GSVNKMVHKP SGQIMAVKRI RSTVDEKEQK QLLMDLDVVM RSSDCPYIVQ FYGALFREGD CWICMELMST SFDKFYKYVY SVLDDVIPEE ILGKITLATV KALNHLKENL KIIHRDIKPS NILLDRSGNI KLCDFGISGQ LVDSIAKTRD AGCRPYMAPE RIDPSASRQG YDVRSDVWSL GITLYELATG RFPYPKWNSV FDQLTQVVKG DPPQLSNSEE REFSPSFINF VNLCLTKDES KRPKYKELLK HPFILMYEER TVEVACYVCK ILDQMPATPS SPMYVD | |
Greater than 90% as determined by SDS-PAGE. |
26398 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse MKK4 / MEK4 / MAP2K4 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction