missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit beta-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7346-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068350

  • 5015.00 DKK / 100µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_058652.1
Hemoglobin subunit beta-2 Protein
Research Use Only
Hbb-b2
Recombinant Protein
E. coli
VHLTDAEKSA VSCLWAKVNP DEVGGEALGR LLVVYPWTQR YFDSFGDLSS ASAIMGNPKV KAHGKKVITA FNEGLKNLDN LKGTFASLSE LHCDKLHVDP ENFRLLGNAI VIVLGHHLGK DFTPAAQAAF QKVVAGVATA LAHKYH
Greater than 90% as determined by SDS-PAGE.
15130
100 μg
Not determined.
Full Length
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers