missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6146-ec-1mg
This item is not returnable.
View return policy
Specifications
Hemoglobin subunit alpha Protein | |
Research Use Only | |
Hba | |
Recombinant Protein | |
E. coli | |
VLSGEDKSNI KAAWGKIGGH GAEYGAEALE RMFASFPTTK TYFPHFDVSH GSAQVKGHGK KVADALASAA GHLDDLPGAL SALSDLHAHK LRVDPVNFKL LSHCLLVTLA SHHPADFTPA VHASLDKFLA SVSTVLTSKY R | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction